Furan-2-boronic acid 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

359390010 359390100
ACRO359390010EA 32.8 GBP
ACRO359390010 ACRO359390100
Furan-2-boronic acid 97%
Furan-2-boronic acid
Formula: C₄H₅BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00799544
CAS Number: 13331-23-2

Order Now

Specification Test Results

Appearance Light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR