5-Methyl-2-furanmethanol 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (5-Methylfuran-2-yl)methanol, 2-(Hydroxymethyl)-5-methylfuran

358510050
ACRO358510050EA 89.8 GBP
ACRO358510050
5-Methyl-2-furanmethanol 97%
5-Methyl-2-furanmethanol

Appearance: Clear yellow Liquid

Formula: C₆H₈O₂
MW: 112.128 g/mol
Boiling Pt: 80…80.5 °C (123 mmHg)
Density: 1.0769 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00130134
CAS Number: 3857-25-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR