Tetrabutylammonium tetrafluoroborate 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Tetrabutylammoniumtetrafluoroborate

353090250 353091000 353090050
ACRO353090250EA 75 GBP
ACRO353090250 ACRO353091000 ACRO353090050
Tetrabutylammonium tetrafluoroborate 98%
Tetrabutylammonium tetrafluoroborate
Formula: C₁₆H₃₆BF₄N
MW: 329.27 g/mol
Melting Pt: 156…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00011634
CAS Number: 429-42-5
EINECS: 207-058-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Assay ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR