Tetrabutylammonium hexafluorophosphate 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Tetrabutylammonium hexafluorophosphate(V), Tetra-n-butylammonium hexafluorophosphate

352580250 352581000 352585000
ACRO352580250EA 50.2 GBP
ACRO352580250 ACRO352581000 ACRO352585000
Tetrabutylammonium hexafluorophosphate 98%
Tetrabutylammonium hexafluorophosphate
Formula: C₁₆H₃₆F₆NP
Melting Pt: 244…246 °C
Storage Temperature: Ambient
MDL Number: MFCD00011748
CAS Number: 3109-63-5
EINECS: 221-472-6
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 242 to 247 °C
Elemental Analysis ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR