Hydrocortisone 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

352450050 352450010 352450250 352451000
ACRO352450050EA 92.4 GBP
ACRO352450050 ACRO352450010 ACRO352450250 ACRO352451000
Hydrocortisone 98%
Hydrocortisone
Formula: C₂₁H₃₀O₅
MW: 362.47 g/mol
Boiling Pt: 567
Melting Pt: 212…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00011654
CAS Number: 50-23-7
EINECS: 200-020-1

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 212 to 217 °C
HPLC ≥97.5 %
Specific optical rotation +150° to +156° (20 °C, 589 nm) (C=1, dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR