Tetramethylammonium hydrogen sulphate 99+% for HPLC

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

343830050 343830500
ACRO343830050EA 86.2 GBP
ACRO343830050 ACRO343830500
Tetramethylammonium hydrogen sulphate 99+% for HPLC
Tetramethylammonium hydrogen sulphate
Formula: C₄H₁₃NO₄S
Melting Pt: 295…297 °C
Storage Temperature: Ambient
MDL Number: MFCD00036149
CAS Number: 80526-82-5
EINECS: 279-490-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
UV (0.005 M in water)
Titration with NaOH ≥99.0 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR