Captopril 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (S)-(-)-1-(3-Mercapto-2-methyl-1-oxopropyl)-L-proline, (S)-(-)-1-(3-Mercapto-2-methylpropionyl)-L-proline, (S)-1-((S)-3-mercapto-2-methylpropanoyl)pyrrolidine-2-carboxylic acid

343700010 343700050 343700250
ACRO343700010EA 43 GBP
ACRO343700010 ACRO343700050 ACRO343700250
Captopril 98%
Captopril

Appearance: White Crystalline powder

Formula: C₉H₁₅NO₃S
Melting Pt: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00168073
CAS Number: 62571-86-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 103 to 108 °C
HPLC ≥97.5 %
Specific optical rotation -125° to -134° (20 °C, 589 nm) (c=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR