Phenosafranine

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3,7-Diamino-5-phenylphenazinium chloride

339340010 339340050
ACRO339340010EA 40.8 GBP
ACRO339340010 ACRO339340050
Phenosafranine
Phenosafranine
Formula: C₁₈H₁₅ClN₄
MW: 322.8 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
CAS Number: 81-93-6
Merck Index: 12,07402

Order Now

Specification Test Results

Appearance Green Crystalline powder
Infrared spectrum Conforms
Dye content ≥80 %
Absorptivity (1%, 1cm) ≥1000 (1 % in water, 517-521 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR