(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

337630010 337630050 337630250
ACRO337630010EA 31.3 GBP
ACRO337630010 ACRO337630050 ACRO337630250
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone 99%
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone
Formula: C₁₀H₁₁NO₂
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00010845
CAS Number: 77943-39-6

Order Now

Specification Test Results

Appearance White Powder or crystals or flakes
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +165° to +175° (20°C, 589 nm) (c=2, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR