(+)-Diacetyl-L-tartaric anhydride 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (3R,4R)-2,5-Dioxotetrahydrofuran-3,4-diyl diacetate

336040050 336040250 336041000
ACRO336040050EA 22.1 GBP
ACRO336040050 ACRO336040250 ACRO336041000
(+)-Diacetyl-L-tartaric anhydride 97%
(+)-Diacetyl-L-tartaric anhydride

CAS No.: 6283-74-5

Formula: C₈H₈O₇
MW: 216.15 g/mol
Melting Pt: 130…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037918
CAS Number: 6283-74-5

Order Now

Specification Test Results

Appearance White Powder or crystals or needles
Infrared spectrum Conforms
Melting point 130 to 135 °C
Specific optical rotation +56° to +62° (20 °C, 589 nm) (c=6, acetone)
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR