Acrylamide : Bisacrylamide (29:1) 40% mix in water for biochemistry

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

330211000 330215000
ACRO330211000EA 42.7 GBP
ACRO330211000 ACRO330215000
Acrylamide : Bisacrylamide (29:1) 40% mix in water for biochemistry
Acrylamide-Bis (32:1)
Density: 1.035 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00080848
CAS Number: 79-06-1
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Solution
UV at 290 nm A: ≤0.1
Acrylic acid (HPLC) ≤0.001 %
Conductivity ≤3 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR