D-(+)-Maltose monohydrate 92+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate

329911000 329915000 329910025
ACRO329911000EA 26.7 GBP
ACRO329911000 ACRO329915000 ACRO329910025
D-(+)-Maltose monohydrate 92+%
D(+)-Maltose monohydrate
Formula: C₁₂H₂₂O₁₁·1H₂O
Melting Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00149343
CAS Number: 6363-53-7
EINECS: 200-716-5
Merck Index: 13,05736
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f861d7adf53134a0c485b5942d3be34c4d0b571&vid=16e5c4425a7a8e158a4200470a45483c7ff5781e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤7 %
Sulfated ash ≤0.1 %
Additional info remainder mainly glucose and maltotriose
HPLC ≥92.0 %
Specific optical rotation on dry substance


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR