CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid) 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid

327741000 327740010 327740250
ACRO327741000EA 71.1 GBP
ACRO327741000 ACRO327740010 ACRO327740250
CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid) 99%
CAPSO (3-(Cyclohexylamino)-2-hydroxypropanesulphonic acid)
Formula: C₉H₁₉NO₄S
MW: 237.32 g/mol
Melting Pt: 263…271 °C
Storage Temperature: Ambient
MDL Number: MFCD00041778
CAS Number: 73463-39-5

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Solubility (0.1M in water) Clear colorless solution
UV at 280 nm A: ≤0.03
pH 5 to 6.5 (1 % in water at 25°C)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR