Petroleum spirit, 40…60 °C for residue analysis, suitable for PAH analysis

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

326790010 326790025
ACRO326790010EA 28.9 GBP
ACRO326790010 ACRO326790025
Petroleum spirit, 40…60 °C for residue analysis, suitable for PAH analysis
Petroleum spirit
Boiling Pt: 40…60 °C (1013 hPa)
Density: 0.65 g/cm³
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.01 % (Coulometric)
Boiling point 40 to 60 °C
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR