Potassium cyanide 97% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

316455000 316450250 316451000 316450025
ACRO316455000EA 136 GBP
ACRO316455000 ACRO316450250 ACRO316451000 ACRO316450025
Potassium cyanide 97% ACS
Potassium cyanide
Formula: KCN
Boiling Pt: 1625 °C
Melting Pt: 634 °C
Density: 1,55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011397
CAS Number: 151-50-8
EINECS: 205-792-3
UN: 1680
ADR: 6.1,I
Merck Index: 13,07709

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Argentometric ≥96.0 %
Chloride (Cl) ≤0.5 %
Sulfide (S) ≤0.003 %
Thiocyanate (SCN) Passes test (Limit about 0.02%)
Phosphate (PO4) ≤0.005 %
Sulfate (SO4) ≤0.04 %
Lead (Pb) ≤2 ppm
Sodium (Na) ≤0.5 %
Iron, total (as Fe) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR