Tellurium 99.8% (trace metals basis), powder -200 mesh

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

315991000 315990250 315995000
ACRO315991000EA 82.8 GBP
ACRO315991000 ACRO315990250 ACRO315995000
Tellurium 99.8% (trace metals basis), powder -200 mesh
Tellurium
Formula: Te
Boiling Pt: 990 °C (760 mmHg)
Melting Pt: 450 °C
Density: 8,83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
ADR: 6.1,III
Merck Index: 12,09273

Order Now

Specification Test Results

Appearance Grey to black Powder
Total trace metal impurities ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR