Digadolinium trioxide 99.9% (trace metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Gadolinium (III) oxide

315510250 315510100 315511000
ACRO315510250EA 65.2 GBP
ACRO315510250 ACRO315510100 ACRO315511000
Digadolinium trioxide 99.9% (trace metals basis)
Digadolinium trioxide
Formula: Gd₂O₃
Melting Pt: 1900 °C
Density: 7,407 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011026
CAS Number: 12064-62-9
EINECS: 235-060-9

Order Now

Specification Test Results

Appearance White Powder
Total conc. of imp.as REO ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR