Tris(4-fluorophenyl)phosphine 98+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

306340010 306340050
ACRO306340010EA 30.3 GBP
ACRO306340010 ACRO306340050
Tris(4-fluorophenyl)phosphine 98+%
Tris(4-fluorophenyl)phosphine
Formula: C₁₈H₁₂F₃P
MW: 316.26 g/mol
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00013553
CAS Number: 18437-78-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 79 to 83 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR