IPTG (isopropyl-β-D-thiogalactopyranoside) 99% for biochemistry dioxane-free

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

302790010 302790100 302790250
ACRO302790010EA 39.2 GBP
ACRO302790010 ACRO302790100 ACRO302790250
IPTG (isopropyl-β-D-thiogalactopyranoside) 99% for biochemistry dioxane-free
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Formula: C₉H₁₈O₅S
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0

Order Now

Specification Test Results

Appearance White Powder or flakes
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Solubility (5 % in water) Clear colorless
HPLC ≥98.5 %
Specific optical rotation -29° to -33° (20°C, 589 nm) (c=1, water)
Dioxane ≤0.1 % (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR