Methanol-[D₃] 99.5 atom % D for NMR spectroscopy

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Carbinol-D3, Hydroxy methane-D3, Methanol-[²H₃], Wood alcohol-D3, Methyl alcohol-D3

300730025 300730100
ACRO300730025EA 66.5 GBP
ACRO300730025 ACRO300730100
Methanol-[D₃] 99.5 atom % D for NMR spectroscopy
Methanol-[D3]
Formula: D₃COH
MW: 35.02 g/mol
Boiling Pt: 65 °C (760 mmHg)
Melting Pt: -98
Density: 0.867 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00055877
CAS Number: 1849-29-2
EINECS: 217-435-9
UN: 1230
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.1 % (Coulometric)
D-Enrichment ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR