(2-Hydroxylpropyl)-β-cyclodextrine 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD

297560250 297561000 297565000
ACRO297560250EA 80.7 GBP
ACRO297560250 ACRO297561000 ACRO297565000
(2-Hydroxylpropyl)-β-cyclodextrine 97%
(2-Hydroxylpropyl)-β-cyclodextrine
Formula: C₆₃H₁₁₂O₄₂
Melting Pt: 278 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD16621721
CAS Number: 128446-35-5

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤7.5 % (K.F.)
HPLC ≤2.0 % Residual beta-cyclodextrin (On dry substance)
NMR substituents are located mainly at O(2) of the anhydroglucopyranose units

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR