Formaldehyde diethyl acetal 99% stabilised, pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ethylal, Diethoxymethane

296450050 296455000
ACRO296450050EA 13.4 GBP
ACRO296450050 ACRO296455000
Formaldehyde diethyl acetal 99% stabilised, pure
Formaldehyde diethyl acetal
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 88
Melting Pt: –66.5 °C
Density: 0.831 g/cm³
Flash Pt: –5 °C
Storage Temperature: Ambient
MDL Number: MFCD00009246
CAS Number: 462-95-3
EINECS: 207-330-6
UN: 2373
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3720 to 1.3760 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer 50 to 150 ppm (BHT)
Peroxides ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR