1,4,7-Triazacyclononane trihydrochloride 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1,4,7-Triazonane trihydrochloride

294315000 294310025
ACRO294315000EA 92.8 GBP
ACRO294315000 ACRO294310025
1,4,7-Triazacyclononane trihydrochloride 97%
1,4,7-Triazacyclononane trihydrochloride
Formula: C₆H₁₈Cl₃N₃
MW: 238.5900 g/mol
Melting Pt: 285…290 °C
Storage Temperature: Ambient
MDL Number: MFCD00074887
CAS Number: 58966-93-1
UN: 3259
ADR: 8 II

Order Now

Specification Test Results

Appearance Off-white to yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR