Palladium 5% on alumina, dry, powder for use as a catalyst

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

293030100 293030500
ACRO293030100EA 150 GBP
ACRO293030100 ACRO293030500
Palladium 5% on alumina, dry, powder for use as a catalyst
Palladium
Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03613602
CAS Number: 7440-05-3

Order Now

Specification Test Results

Appearance Silver grey to black Powder
Assay 4.0 to 6.0 % (Pd)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR