Cesium carbonate 99.99% (trace metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

278020100 278020500
ACRO278020100EA 33.3 GBP
ACRO278020100 ACRO278020500
Cesium carbonate 99.99% (trace metals basis)
Cesium carbonate
Formula: Cs₂CO₃
MW: 325.82 g/mol
Melting Pt: >400 °C
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9
Merck Index: 13,02020

Order Now

Specification Test Results

Appearance White Powder
Total trace metal impurities ≤0.01 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR