Triisopropanolamine cyclic borate 98+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3,7,10-Trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo[3.3.3]undecane, Triisopropanolamine Borate, Triisopropanolamine Cyclic Borate

269241000 269245000
ACRO269241000EA 94.4 GBP
ACRO269241000 ACRO269245000
Triisopropanolamine cyclic borate 98+%
Triisopropanolamine cyclic borate

CAS No.: 101-00-8

Formula: C₉H₁₈BNO₃
Melting Pt: 150…157 °C
Storage Temperature: Ambient
MDL Number: MFCD03791298
CAS Number: 101-00-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline needles and/or chunks
Infrared spectrum Conforms
Melting point 150 to 157 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR