Tetrahydrofuran 99.8% unstabilised for HPLC

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

268290010 268290025 268295000
ACRO268290010EA 49.7 GBP
ACRO268290010 ACRO268290025 ACRO268295000
Tetrahydrofuran 99.8% unstabilised for HPLC
Tetrahydrofuran
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: –108.4 °C
Density: 0.88 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 %
GC ≥99.8 %
UV at 280 nm A: ≤0.02
Wavelength at 1 A.U. ≤215 nm
Residue after evaporation ≤2 ppm
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0006 meq/g
Filtered to 0.2 micrometer Filtered
Peroxides ≤0.0075 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR