1-Chlorododecane 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

257102500 257100010
ACRO257102500EA 25.5 GBP
ACRO257102500 ACRO257100010
1-Chlorododecane 99%
1-Chlorododecane
Formula: C₁₂H₂₅Cl
Boiling Pt: 260 °C (760 mmHg)
Melting Pt: –9.3 °C
Density: 0.87 g/cm³
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00013688
CAS Number: 112-52-7
EINECS: 203-981-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4420 to 1.4440 (20°C, 589 nm)
GC ≥98.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR