Hydroxypropyl methylcellulose (Hypromellose), M.W. 86,000, 4000 cP (2% H₂O)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Methocel K100, Hydroxypropyl methyl cellulose

244021000 244022500 244020010
ACRO244021000EA 128 GBP
ACRO244021000 ACRO244022500 ACRO244020010
Hydroxypropyl methylcellulose (Hypromellose), M.W. 86,000, 4000 cP (2% H₂O)
Hydroxypropyl methyl cellulose (Hypromellose)
Formula: C₃H₈O₂·xCH₄O
Density: 1.39 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00131360
CAS Number: 9004-65-3

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Loss on drying ≤10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR