Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Benzo-18-crown 6-Ether

243980010 243980050
ACRO243980010EA 40.3 GBP
ACRO243980010 ACRO243980050
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 97%
Benzo-18-crown-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecane)
Formula: C₁₆H₂₄O₆
MW: 312.36 g/mol
Melting Pt: 40…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00062741
CAS Number: 14098-24-9

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 40 to 45 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR