Polysorbate 20

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated

233360010 233360050 233362500 233360051 233360250
ACRO233360010EA 63.1 GBP
ACRO233360010 ACRO233360050 ACRO233362500 ACRO233360051 ACRO233360250
Polysorbate 20
Polysorbate 20
Formula: C₅₈H₁₁₄O₂₆
Boiling Pt: >100 °C (760 mmHg)
Density: 1.1 g/cm³
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now

Specification Test Results

Appearance Clear to slightly hazy yellow to brown-yellow Viscous liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.4680 to 1.4720 (20°C, 589 nm)
Water ≤3 %
Dioxane ≤10 ppm
Acid value ≤2 mg KOH/g
Hydroxyl value 96 to 108 mg KOH/g
Saponification value 40 to 50 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR