2,3-Dichlorobenzaldehyde 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

232670250 232671000
ACRO232670250EA 39.6 GBP
ACRO232670250 ACRO232671000
2,3-Dichlorobenzaldehyde 99%
2,3-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
Boiling Pt: 144 °C (30 mmHg)
Melting Pt: 60…64 °C
Density: 1,33 g/cm³ (100 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00010127
CAS Number: 6334-18-5
EINECS: 228-711-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 60 to 64 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR