1-Propanol 99.5% for analysis

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

232070010 232070025 232070100
ACRO232070010EA 44 GBP
ACRO232070010 ACRO232070025 ACRO232070100
1-Propanol 99.5% for analysis
1-Propanol
Formula: H₃CCH₂CH₂OH
Boiling Pt: 97 °C (760 mmHg)
Melting Pt: –127 °C
Density: 0.8 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 %
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Calcium (Ca) ≤0.5 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Aldehyde (C3H6O) ≤0.01 % (GC)
Acetone ≤0.01 % (GC)
Ethanol ≤0.05 % (GC)
Methanol ≤0.05 % (GC)
2-Propanol ≤0.05 % (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR