γ-Cyclodextrine hydrate 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: γ-Cyclodextrin hydrate, γ-Cyclodextrin xhydrate

229900010 229905000 229900050
ACRO229900010EA 168 GBP
ACRO229900010 ACRO229905000 ACRO229900050
γ-Cyclodextrine hydrate 99%
γ-Cyclodextrine hydrate
Formula: C₄₈H₈₀O₄₀·H₂O
MW: 1297.15 g/mol (anhy)
Melting Pt: 267 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149574
CAS Number: 91464-90-3
EINECS: 241-482-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤11 %
HPLC ≥98.5 % (ELSD)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR