L(+)-Potassium hydrogen tartrate 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Potassium hydrogen L-tartrate, Potassium acid tartrate, Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Potassium Bitartrate, (R,R)-(+)-Tartaric acid potassium salt, L(+)-Tartaric acid potassium salt

222670010 222670051 222672500
ACRO222670010EA 44.7 GBP
ACRO222670010 ACRO222670051 ACRO222672500
L(+)-Potassium hydrogen tartrate 99%
L(+)-Potassium hydrogen tartrate
Formula: KOOC(CHOH)₂COOH
Melting Pt: 250 °C
Density: 1.95 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00065392
CAS Number: 868-14-4
EINECS: 212-769-1
Merck Index: 13,07697
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=355f72f319f5589f12d60c6dfffecdec31cee083&vid=4a750ad5d4a91fd4c2885832ee80b7b64cdcc42a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to white Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C)
Specific optical rotation +8.0° to +9.2° (20°C, 589 nm) (c=10, HCl 1M and H2O) (on dried substance)
Titration with NaOH 98.5 to 101.5 % (On dried substance)
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR