4-Methyl-2-pentanone 99.5%, Spectrophotometric Grade

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK

222170025 222175000 222170010
ACRO222170025EA 104 GBP
ACRO222170025 ACRO222175000 ACRO222170010
4-Methyl-2-pentanone 99.5%, Spectrophotometric Grade
4-Methyl-2-pentanone
Formula: (CH₃)₂CHCH₂COCH₃
MW: 100.16 g/mol
Boiling Pt: 117.4 °C (760 mmHg)
Melting Pt: –84 °C
Density: 0.8 g/cm³
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3 II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 %
GC ≥99.5 %
UV at 400 nm A: ≤0.01
Color scale ≤15 APHA
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR