Chloroiodomethane 98% stabilised

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

222150250 222151000 222155000
ACRO222150250EA 63.6 GBP
ACRO222150250 ACRO222151000 ACRO222155000
Chloroiodomethane 98% stabilised
Chloroiodomethane
Formula: CH₂ClI
MW: 176.38 g/mol
Boiling Pt: 108…109 °C (760 mmHg)
Density: 2.42 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00001078
CAS Number: 593-71-5
EINECS: 209-804-8
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to pink Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5810 to 1.5840 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer Copper


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR