Chlorobenzene 99+% for spectroscopy

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

222115000 222110025
ACRO222115000EA 41.3 GBP
ACRO222115000 ACRO222110025
Chlorobenzene 99+% for spectroscopy
Chlorobenzene
Formula: C₆H₅Cl
MW: 112.56 g/mol
Boiling Pt: 131 °C (760 mmHg)
Melting Pt: –45 °C
Density: 1.1075 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3 III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
GC ≥99.0 %
UV at 360 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR