Allyl chloroformate 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

221740500 221741000
ACRO221740500EA 98.9 GBP
ACRO221740500 ACRO221741000
Allyl chloroformate 97%
Allyl chloroformate
Formula: C₄H₅ClO₂
MW: 120.54 g/mol
Boiling Pt: 109…110 °C (760 mmHg)
Density: 1.13 g/cm³
Flash Pt: 31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000648
CAS Number: 2937-50-0
EINECS: 220-916-6
UN: 1722
ADR: 6.1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4210 to 1.4240 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR