1,1,3,3-Tetramethylguanidine 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N,N,N',N'-Tetramethylguanidine

221291000 221295000 221290020
ACRO221291000EA 32 GBP
ACRO221291000 ACRO221295000 ACRO221290020
1,1,3,3-Tetramethylguanidine 99%
1,1,3,3-Tetramethylguanidine
Formula: C₅H₁₃N₃
MW: 115.18 g/mol
Boiling Pt: 160…162 °C (760 mmHg)
Melting Pt: –30 °C
Density: 0.91 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00008323
CAS Number: 80-70-6
EINECS: 201-302-7
UN: 2920
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4665 to 1.4695 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR