(R)-(+)-1-(1-Naphthyl)ethylamine 99+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

217800010 217800050
ACRO217800010EA 36.1 GBP
ACRO217800010 ACRO217800050
(R)-(+)-1-(1-Naphthyl)ethylamine 99+%
(R)-(+)-1-(1-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
Boiling Pt: 153 °C (11 mmHg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00064114
CAS Number: 3886-70-2
EINECS: 223-425-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.6215 to 1.6245 (20°C, 589 nm)
GC ≥99.0 %
Specific optical rotation +51° to +59° (20°C, 589 nm) (c=2, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR