4'-Ethoxyacetanilide 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide

216591000 216595000 216590025
ACRO216591000EA 27.9 GBP
ACRO216591000 ACRO216595000 ACRO216590025
4'-Ethoxyacetanilide 97%
4'-Ethoxyacetanilide
Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Melting Pt: 133…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00009094
CAS Number: 62-44-2
EINECS: 200-533-0

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 133 to 138 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR