O-Phenyl chlorothioformate 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

215490050 215490010 215490250
ACRO215490050EA 65.6 GBP
ACRO215490050 ACRO215490010 ACRO215490250
O-Phenyl chlorothioformate 99%
O-Phenyl chlorothioformate
Formula: C₇H₅ClOS
Boiling Pt: 81…83 °C (6 mmHg)
Density: 1.248 g/cm³
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004920
CAS Number: 1005-56-7
EINECS: 213-736-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5800 to 1.5830 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR