Vanadyl sulphate pentahydrate 17-23% (v/v)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Basic vanadium (IV) sulphate pentahydrate, Vanadium oxide sulphate pentahydrate, Vanadium (IV) sulphate oxide pentahydrate

214841000 214840250
ACRO214841000EA 58.6 GBP
ACRO214841000 ACRO214840250
Vanadyl sulphate pentahydrate 17-23% (v/v)
Vanadyl sulphate pentahydrate
Formula: VOSO₄·5H₂O
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00149933
CAS Number: 12439-96-2
EINECS: 248-652-7
UN: 2931
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Blue Moist crystalline powder or crystals
Degree of hydration ca. 5 moles water
Titration with KMnO4 17 to 23 % (V)
Iron (Fe) ≤600 ppm
Potassium (K) ≤500 ppm
Sodium (Na) ≤500 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR