, powder <50 µm

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

214795000 214790025
ACRO214795000EA 56.3 GBP
ACRO214795000 ACRO214790025
, powder <50 µm
Molecular sieve 3A (0.3 nm, 3 Å)
Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now

Specification Test Results

Appearance White Powder
Particle size ≤50 µm
pH ≤11.4 (10 % suspension in water)
Residual water ≤3 % (550°C, 2 h)
Water adsorption ≥18 % (60% rH, 25°C, 24h)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR