Tetrabutylammonium hydroxide 1 M in methanol

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

212911000 212918000 212910025
ACRO212911000EA 46.9 GBP
ACRO212911000 ACRO212918000 ACRO212910025
Tetrabutylammonium hydroxide 1 M in methanol
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Melting Pt: –98 °C
Density: 0.83 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3286
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution
Titration with HCl 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR