Iodine 99.5+% analysis_for

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

212490250 212491000 212495000
ACRO212490250EA 19.9 GBP
ACRO212490250 ACRO212491000 ACRO212495000
Iodine 99.5+% analysis_for
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 185 °C (760 mmHg)
Melting Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 8 III
Merck Index: 13,05036

Order Now

Specification Test Results

Appearance Dark blue to black Glistening spheres
Titration Iodometric 99.5 to 100.5 %
Chlorine and Bromine (as Cl) ≤0.005 %
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤5 ppm
Magnesium (Mg) ≤5 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤50 ppm
Zinc (Zn) ≤5 ppm
Insoluble matter Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR