Copper(I) chloride 99.99% (trace metals basis), extra pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Cuprous chloride, Copper chloride

212420100 212421000
ACRO212420100EA 74.3 GBP
ACRO212420100 ACRO212421000
Copper(I) chloride 99.99% (trace metals basis), extra pure
Copper (I) chloride
Formula: ClCu
Boiling Pt: 1367 °C (1013 hPa)
Melting Pt: 430 °C
Density: 4.14 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8,III
Merck Index: 13,02690

Order Now

Specification Test Results

Appearance White to light green Crystalline powder
Total trace metal impurities ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR