Hydrazine hydrate 80% (∼51% w/w N₂H₄)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

209592500 209590010
ACRO209592500EA 30.8 GBP
ACRO209592500 ACRO209590010
Hydrazine hydrate 80% (∼51% w/w N₂H₄)
Hydrazine hydrate
Formula: NH₂NH₂·xH₂O
Melting Pt: –57 °C
Density: 1.028 g/cm³
Flash Pt: 91 °C
MDL Number: MFCD00149931
CAS Number: 10217-52-4
EINECS: 206-114-9
UN: 2030
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Refractive index 1.4110 to 1.4140 (20°C, 589 nm)
Density (20°C) 1.027 to 1.030 g/ml
Titration with HCl 78.9 to 80.5 % (N2H4.H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR