Copper(I) chloride 99%, extra pure, purified

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Cuprous chloride, Copper chloride

208390250 208391000 208395000
ACRO208390250EA 35.9 GBP
ACRO208390250 ACRO208391000 ACRO208395000
Copper(I) chloride 99%, extra pure, purified
Copper (I) chloride
Formula: ClCu
Boiling Pt: 1490 °C (760 mmHg)
Melting Pt: 430 °C
Density: 4.14 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8,III
Merck Index: 13,02690

Order Now

Specification Test Results

Appearance Light green to grey Crystalline powder
Titration Complexometric 98.5 to 101.5 %
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤200 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR